Bottle filling bells, Nalgene™
Lieferant: Thermo Fisher Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
DS0390-0070
513-3901EA 61.6 EUR InStock
513-3901
Bottle filling bells, Nalgene™
Flaschen Zellkulturflaschen
Transparent, autoclavable filling bell, made of PC with PP tubing adapter, is for use in transferring sterile media aseptically from large containers to media bottles or culture vessels in microbiology and cell culture labs.
  • Autoclavable polypropylene tubing adapter accepts 6,3, 9,5 and 12,7 mm Int. Ø tubing
  • Bell is durable, non-toxic and will not shatter
  • Costs much less than glass
Note: For research use only; not for in vitro diagnosis or parenterals.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge