Schadstoffmonitore
Lieferant: 3M
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
3551
514-0007EA 448 EUR InStock
514-0007
Schadstoffmonitore
Gas-Spürsysteme
Monitor uses the proven scientific principle of diffusion which involves the gradual mixing of molecules of two or more substances for measuring personal exposure in work areas of industries.
  • Ethylene oxide monitor utilises brominated carbon to collect and convert the reactive ethylene oxide to a more stable chemical derivative, 2-bromoethanol
  • Compound then desorbed with methylene chloride in methanol
  • Accuracy within ±25% for 8-hour samplings at 1 ppm
  • Accuracy within ±35% for 8-hour samplings at the action limit
Haltbarkeit 18 Monate.
Zertifizierungen: OSHA
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge