Multi-element standard solution 5 for ICP, TraceCERT®, Supelco®
Lieferant: SUPELCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
54704-100ML
54704-100MLEA 172 EUR InStock
54704-100ML
Multi-element standard solution 5 for ICP, TraceCERT®, Supelco®
Standards Kationischer Standards Mehrelement-Standards
27 components:
Ag, 10 mg/l; Al, 10 mg/l; Ba, 10 mg/l; Be, 10 mg/l; Bi, 10 mg/l
Ca, 100 mg/l; Cd, 10 mg/l; Co, 10 mg/l; Cr, 10 mg/l; Cs, 10 mg/l; Cu, 10 mg/L
Fe, 100 mg/l; Ga, 10 mg/l; In, 10 mg/l; K, 100 mg/l; Li, 10 mg/l
Mg, 10 mg/l; Mn, 10 mg/l; Mo, 10 mg/l; Na, 100 mg/l; Ni, 10 mg/l
Pb, 10 mg/l; Rb, 10 mg/l; Sr, 10 mg/l; Tl, 10 mg/l; V, 10 mg/l; Zn, 10 mg/l
Zertifizierungen: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge