GC septa, TR-Blue
Lieferant: Thermo Fisher Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
313B3233 313B3240
555-8258EA 45.5 EUR InStock
555-8258 555-8259
GC septa, TR-Blue
Septen
Made of low-bleed silicone, these septa have excellent mechanical properties and are ideal for demanding GC-MS applications.
  • General purpose septa
  • Easy to penetrate
  • Maximum temperature 200 to 250 °C
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge