Solid phase extraction disk, ENVI™-18
Lieferant: Sigma-Aldrich
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
57171
57171.EA 259 EUR InStock
57171.
Solid phase extraction disk, ENVI™-18
Filter Filterscheiben
Solid phase extraction disks provides faster flow rates and exhibits less clogging than PTFE discs for the extraction of organic contaminants from drinking water samples.
  • Reversed phase retention mechanism
  • Porous glass fiber membranes embedded with C18 or C8 modified silica particles
Applications include polynuclear aromatic hydrocarbons (PAHs), polychlorinated biphenyls (PCBs), phthalates, semivolatile organics, paraquat and diquat, pesticides and herbicides.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge