VWR® TraceableLive®, CO₂, Humidity and Temperature Data Logger
Lieferant: VWR Collection
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
620-2694EA 1030 EUR InStock
620-2694
VWR® TraceableLive®, CO₂, Humidity and Temperature Data Logger
Datenlogger
TraceableLIVE® provides simple, digital monitoring of CO₂, temperature and humidity in greenhouses, micro labs, biofuel research and other environments. One free user license in standard delivery.
  • Ambient unit with dongle - place entire unit inside a greenhouse or other environments
  • Cable probe is ideal for CO₂ incubators
  • Alerts for CO₂, temperature, humidity, connectivity interruption and low battery
  • Secure SSL-encrypted data transmission
  • Alarm notifications available via Mobile Push, E-mail and SMS text message
  • Unlimited scalability, multiple point calibration traceable to NIST
  • AC power runs unit, batteries designed as back-up power in case of a power-outage
Probes are ambient dongle sensor, cable sensor and meter cable.
Bestellinformation: A user license for more than one user is available; contact your Avantor representative for more information.
Lieferumfang: Supplied with one ambient external sensor, quick start guide, AC adapter, batteries, magnets, hook and loop tape, wall mount and Traceable® certificate.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge