Handheld refractometer, HHR-2N
Lieferant: ATAGO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
2522
635-0140EA 341 EUR InStock
635-0140
Handheld refractometer, HHR-2N
Refraktometer Handrefraktometer
This refractometer is designed to determine the water content in honey.
  • Scale to measure the percentage of water
  • Refractive index (RI) of sucrose
  • Determination of moisture in honey
Specifically suitable for use in food applications.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge