Kjeldahl distillation system, VAPODEST® 200
Lieferant: GERHARDT
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
12-0520
705-0857EA 0 EUR InStock
705-0857
Kjeldahl distillation system, VAPODEST® 200
Destillationssysteme Kjeldahl-Destillationssysteme
VAPODEST® 200 is the ideal instrument for distillation of volatiles, SO₂, alcohol and phenol.
  • Intelligent supervision of all vital functions
  • Programmable, automatic addition of NaOH
  • Programmable distillation and reaction time
  • Efficient with programmable steam power 10 to 100%
  • 7" TFT fully coloured display and easy menu navigation
VAPODEST® 200 complies with the data management requirements of accredited laboratories (ISO 17025, GLP).
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge