DispoCut™ disposable dissecting boards
Lieferant: CELLPATH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
CGB-0502-53A
720-2330EA 146 EUR InStock
720-2330
DispoCut™ disposable dissecting boards
Präparierbretter
DispoCut™ is a disposable cutting board designed to increase safety and hygiene in high-risk areas as well we routine cutting areas.
  • High-quality laminated board manufactured from 3 mm, three-ply foam for strength and rigidity
  • High degree of moisture resistance
  • Helps protect works surfaces whilst ensuring accurate dissection
Each DispoCut™ board has a metric and imperial scale to assist in the macroscopic and photographic recording of specimens.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge