J.T.Baker®, Multipurpose Alkaline Detergent, RBS 35 concentrate
Lieferant: Avantor
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
7348.5000EA 110 EUR InStock
7348.5000
J.T.Baker®, Multipurpose Alkaline Detergent, RBS 35 concentrate
Reinigungsmittel Waschmittel und Oberflächenreiniger
RBS 35 concentrate is used for manual cleaning, soaking and ultrasonic baths. These surfactants are alkaline, foaming and insensitive to water hardness.
  • High wetting and emulsifying power
  • No risk of calcareous deposits when diluted in tap water
  • Total rinse
Suitable for cleaning glassware, laboratory equipments and for cleaning parts made of glass, stainless steel, injection moulds and production equipment.
Typical concentration for use: 2%
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge