74626-10MG
74626-10MGEA
63.3
EUR
InStock
74626-10MG
Organic reference standard, (E)-2-Methylglutaconic acid, ≥98,0% (HPLC), Supelco®
Standards
Organische Standards
Biochemical and physiol actions are 2-Methylglutaconic acid is found in the urine of patients with organic aciduria from 2-methylacetoacetyl-CoA thiolase.