Organic reference standard, (E)-2-Methylglutaconic acid, ≥98,0% (HPLC), Supelco®
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
74626-10MG
74626-10MGEA 63.3 EUR InStock
74626-10MG
Organic reference standard, (E)-2-Methylglutaconic acid, ≥98,0% (HPLC), Supelco®
Standards Organische Standards
Biochemical and physiol actions are 2-Methylglutaconic acid is found in the urine of patients with organic aciduria from 2-methylacetoacetyl-CoA thiolase.
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f53c3b238ecac67761c986948dde0d70a4c6e12&vid=3f53c3b238ecac67761c986948dde0d70a4c6e12&prid=web&clid=SAPDAM
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge