Dot blot system, Enhancer™
Lieferant: G-Biosciences
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
786-163
786-163EA 2430 EUR InStock
786-163
Dot blot system, Enhancer™
Blotting-Apparate
Enhancer™ dot blot system allows for the rapid application of up to 384 protein or nucleic acid samples on to a 12×9 cm membrane for subsequent analysis. Samples are applied to a nitrocellulose or nylon membrane by capillary action, resulting in concentration of sample due to minimal diffusion.
  • Increase dot blot sensitivity by 10-fold
  • Apply up to 384 protein or nucleic acid samples
  • No cross-contamination risk
  • No expensive electronic equipment required
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge