Organic reference standard, 4-Hydroxy-4-methyl-2-pentanone, analytical standard, Supelco®
Lieferant: SUPELCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
78891-5ML
78891-5MLEA 226 EUR InStock
78891-5ML
Organic reference standard, 4-Hydroxy-4-methyl-2-pentanone, analytical standard, Supelco®
Standards Organische Standards
4-Hydroxy-4-methyl-2-pentanone may be used as an internal standard in the analysis of volatile odor-active compounds present in wine using solid phase extraction with gas chromatography (SPE-GC).
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cf45d35a8390bbc783b8331d25a329a2a75840d4&vid=cf45d35a8390bbc783b8331d25a329a2a75840d4&prid=web&clid=SAPDAM
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge