Blot module, miniVE™
Lieferant: Cytiva
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
80-6418-96
80-6418-96EA 637 EUR InStock
80-6418-96
Blot module, miniVE™
Blotting-Apparate
Perform 1-D and 2-D electrophoresis and electrotransfer in a single compact unit.
  • Complete semi-wet blotting in 45 minutes with only 300 ml buffer
  • Choose precast gels from a variety of manufacturers
  • Perform two second-dimension separations using 7 cm IPG strips in 15 hours
Cast, run, and blot mini-gels in four streamlined components: Gel module, blot module, lower buffer chamber, safety lid.
Two mini-gels or four mini-blots at once, using 100×105 mm gel cassettes for increased separation distances.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge