Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Calcein AM ≥90% (durch HPLC) fluoreszierender Farbstoff, für die Histologie

Lieferant: BIOTIUM

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


80011 80011-3
80011.EA 331 EUR InStock
80011. 80011-3
Calcein AM ≥90% (durch HPLC) fluoreszierender Farbstoff, für die Histologie
Calcein AM
Calcein AM is a widely used green fluorescent cell marker. Calcein AM is membrane-permeant and thus can be introduced into cells via incubation.

  • λEx/λEm (calcein) = 494/517 nm (pH 8)
  • Colorless solid soluble in DMSO
  • Store at -20°C and protect from light

Once inside the cells, calcein AM is hydrolyzed by endogenous esterase into the highly negatively charged green fluorescent calcein, which is retained in the cytoplasm. Calcein AM is an excellent tool for the studies of cell membrane integrity and for cell tracing. Supplied in lyophilized form or as a solution in anhydrous DMSO at 4 mM (80011-1) or 1 mg/mL (1 mM) (80011-2). Calcein AM is also offered in our Cell Viability and Cytotoxicity Assay Kit (30002).


Formel: C₄₆H₄₆N₂O₂₃
Storage Temperature: Gefrierschrank
MDL Number: MFCD05861516
CAS-Nummer: 148504-34-1
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b6608e2f014f82d078dfeecaf9d7e7c8b1e55a83&vid=b6608e2f014f82d078dfeecaf9d7e7c8b1e55a83&prid=web&clid=SAPDAM
Order Now

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge