90047
90047.EA
242
EUR
InStock
90047.
NBD-Ethylenediamine fluoreszierender Farbstoff
NBD-Ethylenediamine
The fluorescence of NBD is highly dependent on the environment; it is virtually nonfluorescent in water but becomes strongly green fluorescent in hydrophobic media. NBD-ethylenediamine can be readily coupled to activated carboxy groups or sulfonyl chlorides.
- λEx/λEm (MeOH) = 466/535 nm
- Yellow solid soluble in DMSO or DMF
- Store at 4°C and protect from light