VWR®, Combined Five Anion Standard, 5 components in H₂O
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91289.180EA 166 EUR InStock
91289.180
VWR®, Combined Five Anion Standard, 5 components in H₂O
Standards Anionische Standards Anionische Multi-Element-Standards
5 components: Fluorides (F-) 20 mg/l, Chlorides (Cl-) 30 mg/l, Nitrates (NO3-) 100 mg/l, Phosphates (PO43-) 150 mg/l, Sulphates (SO42-) 150 mg/l in H₂O
Zertifizierungen: ISO 17025/17034
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge