91289.180EA
166
EUR
InStock
91289.180
VWR®, Combined Five Anion Standard, 5 components in H₂O
Standards
Anionische Standards
Anionische Multi-Element-Standards
5 components: Fluorides (F-) 20 mg/l, Chlorides (Cl-) 30 mg/l, Nitrates (NO3-) 100 mg/l, Phosphates (PO43-) 150 mg/l, Sulphates (SO42-) 150 mg/l in H₂O
Zertifizierungen: ISO 17025/17034