A10263.0E
A10263.22
A10263.36
A10263.0EEA
198
EUR
InStock
A10263.0E
A10263.22
A10263.36
di-Ammoniumoxalat Monohydrat ≥98%
di-Ammoniumoxalat Monohydrat
Purity: 98%
CAS No.: 6009-70-7
Molecular Formula: C2H8N2O4.H2O
Formula Weight: 142.11 (124.10anhy)
Physical Form: Crystalline solid
Density: 1.50
Appearance: White
Melting Point: 70° dec.
Beilstein Reference: 3759830
Merck Reference: 14,537
UN No.: 3288
MDL No.: MFCD00149694