A10276.18
A10276.30
A10276.0B
A10276.18EA
26
EUR
InStock
A10276.18
A10276.30
A10276.0B
Eisen(III)citrat Hydrat Fe(III) 16,5-20% (Fe(II) ≤ 5%)
Eisen(III)citrat Hydrat
Provides a source of iron used in cell culture applications. Provides iron in a less toxic form than free iron salts.
CAS No.: 334024-15-6
Molecular Formula: C6H5FeO7.xH2O
Formula Weight: 244.94(anhy)
MDL No.: MFCD00149617