A10312.14
A10312.22
A10312.36
A10312.14EA
35.2
EUR
InStock
A10312.14
A10312.22
A10312.36
N-Hydroxysuccinimid ≥98%
N-Hydroxysuccinimid
Purity: 98+%
CAS No.: 6066-82-6
Molecular Formula: C4H5NO3
Formula Weight: 115.09
Melting Point: 94-99°
Beilstein Reference: 113913