A10333.09
A10333.18
A10333.09EA
39.9
EUR
InStock
A10333.09
A10333.18
2-(4-Hydroxyphenylazo)benzoesäure ≥98%
2-(4-Hydroxyphenylazo)benzoesäure
Purity: 98+%
CAS No.: 1634-82-8
Molecular Formula: C13H10N2O3
Formula Weight: 242.23
Melting Point: 205-207°
Beilstein Reference: 1842696