A10513.14
A10513.22
A10513.14EA
29
EUR
InStock
A10513.14
A10513.22
2-Chlor-6-fluorbenzaldehyd ≥97%
2-Chloro-6-fluorobenzaldehyde
Purity: 97%
CAS No.: 387-45-1
Molecular Formula: C7H4ClFO
Formula Weight: 158.56
Flash Point: 101°(213°F)
Melting Point: 35-39°
Boiling Point: 91-93°/10mm
Beilstein Reference: 2245530
MDL No.: MFCD00003306