A10760.06
A10760.14
A10760.22
A10760.06EA
43.5
EUR
InStock
A10760.06
A10760.14
A10760.22
Kojisäure ≥99%
Kojisäure
Purity: 99%
CAS No.: 501-30-4
Molecular Formula: C6H6O4
Formula Weight: 142.11
Odour: Odourless
Melting Point: 152-155°
Beilstein Reference: 120895
Merck Reference: 14,5317
MDL No.: MFCD00006580