A11520.0B
A11520.18
A11520.30
A11520.0BEA
656
EUR
InStock
A11520.0B
A11520.18
A11520.30
Ethyltrifluoracetat ≥99%
Ethyltrifluoracetat
Purity: 99%
CAS No.: 383-63-1
Molecular Formula: C4H5F3O2
Formula Weight: 142.08
Flash Point: -1°(30°F)
Refractive Index: 1.3070
Odour: Sweet
Melting Point: -78°
Boiling Point: 60-62°
Beilstein Reference: 1761411
UN No.: 2924
MDL No.: MFCD00000419