A12484.22
A12484.36
A12484.22EA
32
EUR
InStock
A12484.22
A12484.36
Methyl-α-D-glucopyranosid ≥98%
Methyl-α-D-glucopyranosid
Purity: 98+%
CAS No.: 97-30-3
Molecular Formula: C7H14O6
Formula Weight: 194.18
Melting Point: 166-171°
Solubility: Soluble in water. Insoluble in ether
Beilstein Reference: 81568
Merck Reference: 14,6080
Optical Rotation: +159° (c=10 in water)
MDL No.: MFCD00064086