A12784.06
A12784.14
A12784.22
A12784.06EA
21.4
EUR
InStock
A12784.06
A12784.14
A12784.22
α,α,α-Trifluor-m-kresol ≥98%
α,α,α-Trifluor-m-kresol
Purity: 98+%
CAS No.: 98-17-9
Molecular Formula: C7H5F3O
Formula Weight: 162.11
Flash Point: 74°(165°F)
Refractive Index: 1.4580
Odour: Phenol-like
Melting Point: -2°
Boiling Point: 177-178°
Beilstein Reference: 2045663
UN No.: 3265
MDL No.: MFCD00002299