A12882.0B
A12882.18
A12882.30
A12882.PG
A12882.0BEA
363
EUR
InStock
A12882.0B
A12882.18
A12882.30
A12882.PG
Ammoniumcer(IV)nitrat ≥98%
Ammoniumcer(IV)nitrat
Purity: 98+%
CAS No.: 16774-21-3
Molecular Formula: H8CeN8O18
Formula Weight: 548.23
Odour: Pungent
Merck Reference: 14,1992
MDL No.: MFCD00151121