A12959.14
A12959.22
A12959.36
A12959.14EA
52.2
EUR
InStock
A12959.14
A12959.22
A12959.36
1-Adamantancarbonsäure ≥99%
1-Adamantancarbonsäure
Purity: 99%
CAS No.: 828-51-3
Molecular Formula: C11H16O2
Formula Weight: 180.25
Melting Point: 173-176°
Beilstein Reference: 1910637