A13531.0B
A13531.0I
A13531.22
A13531.30
A13531.0BEA
142
EUR
InStock
A13531.0B
A13531.0I
A13531.22
A13531.30
p-Kresol ≥99%
p-Kresol
Purity: 99%
CAS No.: 106-44-5
Molecular Formula: C7H8O
Formula Weight: 108.14
Flash Point: 89°(192°F)
Odour: Phenol-like
Boiling Point: 201-202°
Beilstein Reference: 1305151
Merck Reference: 14,2579
UN No.: 3455
MDL No.: MFCD00002376