A14027.03
A14027.06
A14027.14
A14027.18
A14027.03EA
22.7
EUR
InStock
A14027.03
A14027.06
A14027.14
A14027.18
Phenylpyruvic acid sodium salt monohydrate 98%
Phenylpyruvic acid sodium salt monohydrate
Purity: 98%
CAS No.: 122049-54-1
Molecular Formula: C9H7NaO3.H2O
Formula Weight: 204.16 (186.14anhy)
Storage Temperature: 2°C to 8°C
Melting Point: 300°
Beilstein Reference: 4770978
MDL No.: MFCD00149109