A14113.06
A14113.14
A14113.06EA
36.7
EUR
InStock
A14113.06
A14113.14
2,4-Difluorbenzonitril ≥98%
2,4-Difluorbenzonitril
Purity: 98%
CAS No.: 3939-09-1
Molecular Formula: C7H3F2N
Formula Weight: 139.11
Flash Point: 110°(230°F)
Odour: Almond-like
Boiling Point: 189°
Beilstein Reference: 1940316
UN No.: 3439
MDL No.: MFCD00009826