A14314.14
A14314.06
A14314.22
A14314.14EA
57.3
EUR
InStock
A14314.14
A14314.06
A14314.22
3-(2-Methoxyphenyl)propionsäure ≥98%
3-(2-Methoxyphenyl)propionsäure
Purity: 98+%
CAS No.: 6342-77-4
Molecular Formula: C10H12O3
Formula Weight: 180.20
Melting Point: 85-88°
Beilstein Reference: 2097012