Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Propylencarbonat 99% (durch GC)

Lieferant: ALFA AESAR

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Achtung

A15552.30 A15552.0B A15552.A4 A15552.XL
A15552.30EA 22.7 EUR InStock
A15552.30 A15552.0B A15552.A4 A15552.XL
Propylencarbonat 99% (durch GC)
Propylencarbonat
Propylene carbonate is a polar, aprotic solvent used in paints and coatings and as a high-permittivity component of electrolytes in lithium batteries.

It plays an important role in adhesives, paint strippers, cosmetics, plasticizer, hard surface cleaner, resin cleaner, fiberglass, polyester and polyurethane cleaner. Furthermore, it acts as a cleaner in polyurethane, a carburetor and is used in stereo lithography parts. In electrospray ionization mass spectrometry, it is doped into low surface tension solutions to enhance analyte charging.

Achtung! Bitte beachten: Moisture sensitive. Hygroscopic. Incompatible with acids and bases.


Formel: C₄H₆O₃
Siedepunkt: 240…243 °C
Melting Pt: –49 °C
Dichte: 1,204
Flash-Pt: 132 °C (270 °F)
Storage Temperature: Raumtemperatur
MDL Number: MFCD00005385
CAS-Nummer: 108-32-7
EINECS: 203-572-1
Order Now

 

PRODUKT

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge