A16944.22
A16944.36
A16944.22EA
37.7
EUR
InStock
A16944.22
A16944.36
Xylit ≥99%
Xylit
Purity: 99%
CAS No.: 87-99-0
Molecular Formula: C5H12O5
Formula Weight: 152.15
Odour: Odourless
Melting Point: 92-96°
Beilstein Reference: 1720523
Merck Reference: 14,10085
MDL No.: MFCD00064292