A10914.18
A10914.09
A10914.30
AB110356.0010
A10914.18EA
152
EUR
InStock
A10914.18
A10914.09
A10914.30
ABCRAB110356.0010
4-Brombenzonitril ≥98%
4-Brombenzonitril
Purity: 98+%
CAS No.: 623-00-7
Molecular Formula: C7H4BrN
Formula Weight: 182.03
Flash Point: 130°(266°F)
Melting Point: 111-114°
Boiling Point: 235-237°
Beilstein Reference: 2041518
UN No.: 3439
MDL No.: MFCD00001811