AB126567.0005
ABCRAB126567.0005EA
0
EUR
InStock
ABCRAB126567.0005
(3,4-Dimethoxyphenylthio)essigsäure ≥97%
(3,4-Dimethoxyphenylthio)essigsäure
Purity: 97%
CAS No.: 95735-63-0
Molecular Formula: C10H12O4S
Formula Weight: 228.26
Melting Point: 101-103°
MDL No.: MFCD00052112