KA6700.1KIT
ABNOKA6700.1KITEA
359
EUR
InStock
ABNOKA6700.1KIT
Toxoplasma IgG ELISA kit
Tests
ELISAs
Toxoplasma IgG ELISA Kit is enzyme immunoassay for the quantitative and qualitative measurement of IgG antibodies to Toxoplasma gondii in human serum and plasma.