Human 2B4, Fc Tag (2B4-H5255) is expressed from human 293 cells (HEK293). It contains AA Cys 22 - Arg 221 (Accession NP057466). 1 * 100 G
Lieferant: ACROBIOSYSTEMS
Human 2B4, Fc Tag (2B4-H5255) is expressed from human 293 cells (HEK293). It contains AA Cys 22 - Arg 221 (Accession NP057466). 1 * 100 G
PRODUKT
Erweitert / Kompakt
Human 2B4, Fc Tag (2B4-H5255) is expressed from human 293 cells (HEK293). It contains AA Cys 22 - Arg 221 (Accession NP057466). 1 * 100 µG ACRB2B4-H5255100UG 1 * 100 µG
Hersteller-Artikelnummer:: 2B4-H5255100UG
Lokale Artikelnummer:: ACRB2B4-H5255100UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder