Porcine FCGRTB2M Heterodimer Protein, His TagStrep II Tag (SPR verified) (FCM-P5280) is expressed from human 293 cells (HEK293). It contains AA Asp 16 - Ser 289 (FCGRT) Val 21 - His 118 (B2M) (Accession Q6XAV9-1 (FCGRT) Q07717-1 (B2M)). 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Porcine FCGRTB2M Heterodimer Protein, His TagStrep II Tag (SPR verified) (FCM-P5280) is expressed from human 293 cells (HEK293). It contains AA Asp 16 - Ser 289 (FCGRT) Val 21 - His 118 (B2M) (Accession Q6XAV9-1 (FCGRT) Q07717-1 (B2M)). 1 * 1 mg
PRODUKT
Erweitert / Kompakt
Porcine FCGRTB2M Heterodimer Protein, His TagStrep II Tag (SPR verified) (FCM-P5280) is expressed from human 293 cells (HEK293). It contains AA Asp 16 - Ser 289 (FCGRT) Val 21 - His 118 (B2M) (Accession Q6XAV9-1 (FCGRT) Q07717-1 (B2M)). 1 * 1 mg ACRBFCM-P5280-1MG 1 * 1 mg
Hersteller-Artikelnummer:: FCM-P5280-1MG
Lokale Artikelnummer:: ACRBFCM-P5280-1MG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder