Recombinat Protein,Human Jagged 1 JAG1 Protein, His Tag,Human Jagged 1, His Tag (JA1-H52H9) is expressed from human 293 cells (HEK2 93). It contains AA Gln 34 - Ser 1046 (Accession P78504-1).This protein carries a polyhistidine tag at the C-terminus 1 * 50 G
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Human Jagged 1 JAG1 Protein, His Tag,Human Jagged 1, His Tag (JA1-H52H9) is expressed from human 293 cells (HEK2 93). It contains AA Gln 34 - Ser 1046 (Accession P78504-1).This protein carries a polyhistidine tag at the C-terminus 1 * 50 G
PRODUKT
Erweitert / Kompakt
Recombinat Protein,Human Jagged 1 JAG1 Protein, His Tag,Human Jagged 1, His Tag (JA1-H52H9) is expressed from human 293 cells (HEK2 93). It contains AA Gln 34 - Ser 1046 (Accession P78504-1).This protein carries a polyhistidine tag at the C-terminus 1 * 50 µG ACRBJA1-H52H9-50UG 1 * 50 µG
Hersteller-Artikelnummer:: JA1-H52H9-50UG
Lokale Artikelnummer:: ACRBJA1-H52H9-50UG
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder