D-ɑ-Tocopherylquinon 90-95%
Lieferant: ACROS ORGANICS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
350305000
ACRO350305000EA 522 EUR InStock
ACRO350305000
D-ɑ-Tocopherylquinon 90-95%
D-alpha-Tocopherylquinone
Appearance: Clear amber to red Viscous liquid
Formel: C₂₉H₅₀O₃
Flash-Pt: >110 °C
Storage Temperature: Kühlschrank
MDL Number: MFCD00135657
CAS-Nummer: 7559-04-8
Order Now
Spezifikation Testergebnisse
Appearance Clear amber to red Viscous liquid
Infrared spectrum Conforms
GC ≥90 %
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge