Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SnRK2.2, SnRK2.3, SnRK2.6 Rabbit Polyclonal Antibody

Lieferant: Agrisera

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

AS142783
AGRIAS142783EA 329 EUR InStock
AGRIAS142783
Anti-SnRK2.2, SnRK2.3, SnRK2.6 Rabbit Polyclonal Antibody
Antikörper
SnRK2.2, SnRK2.3, SnRK2.6 are members of SNF1-related protein kinases whose activity is activated by ionic (salt) and non-ionic (mannitol) osmotic stress. Involved in ABA signalling pathway. Alternative names: OST1-kinase-like 2, Protein ATHPROKIN B, SNF1-related kinase 2.3, SnRK2.3, Protein OPEN STOMATA 1SNF1-related kinase 2.6, SnRK2.6, Serine/threonine-protein kinase OST1, OST1-kinase-like 3, Protein ATHPROKIN A, SNF1-related kinase 2.2, SnRK2.2,SnRK2.2, SnRK2.3, SnRK2.6 are members of SNF1-related protein kinases whose activity is activated by ionic (salt) and non-ionic (mannitol) osmotic stress. Involved in ABA signalling pathway. Alternative names: OST1-kinase-like 2, Protein ATHPROKIN B, SNF1-related kinase 2.3, SnRK2.3, Protein OPEN STOMATA 1SNF1-related kinase 2.6, SnRK2.6, Serine/threonine-protein kinase OST1, OST1-kinase-like 3, Protein ATHPROKIN A, SNF1-related kinase 2.2, SnRK2.2

Type: Primary
Antigen: SNRK2.2/2.3/2.6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge