HIGH STAT PLUS ESD Trousers
Lieferant: ALSICO HIGHTECH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
1017450-BE40 1017450-BE42 1017450-BE44 1017450-BE46 1017450-BE48 1017450-BE50 1017450-BE52 1017450-BE54 1017450-BE56 1017450-BE58 1017450-BE60
ALSO1017450-BE40EA 64.8 EUR InStock
ALSO1017450-BE40 ALSO1017450-BE42 ALSO1017450-BE44 ALSO1017450-BE46 ALSO1017450-BE48 ALSO1017450-BE50 ALSO1017450-BE52 ALSO1017450-BE54 ALSO1017450-BE56 ALSO1017450-BE58 ALSO1017450-BE60
HIGH STAT PLUS ESD Trousers
Arbeitskleidung
Unisex ESD trousers.
  • Waistband with 5 belt loops, closed at the front with hidden button
  • 2 back pockets
  • Thigh pocket with flap on left leg
  • Double metre pocket on right leg
  • Accents in royal blue
  • ESD logo
Industrial launderable.
Zertifizierungen: Oeko-Tex® standard 100; IEC 61340 - 4 - 9:2016.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge