Chemical resistant shoe covers, MICROGARD® 2500, model 400
Lieferant: ANSELL HEALTH CARE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
WH25B-00400-00
ANSEWH25B-00400-00EA 999 EUR InStock
ANSEWH25B-00400-00
Chemical resistant shoe covers, MICROGARD® 2500, model 400
Einweg-Kleidung
These over shoes are made of microporous PP film with a non woven inner layer. The fabric provides an excellent barrier to chemical spray and infective agents.
  • Elasticated opening
  • Bound seams
  • Fit shoe size 42 - 46
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge