SOCO boot covers, MICROGARD® 2500, model 409
Lieferant: ANSELL HEALTH CARE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
WH25B-00409-00
ANSEWH25B-00409-00EA 705 EUR InStock
ANSEWH25B-00409-00
SOCO boot covers, MICROGARD® 2500, model 409
Einweg-Kleidung
These SOCO boot protectors are made from a non woven fabric with microporous PP film. The laminate provides an excellent barrier to chemical spray and infective agents. Developed specifically for police forensic Scene of Crime Officers (SOCOs).
  • Elastic to top of boot
  • Tie fastening
  • Blue binding to seams
  • Reinforced Surestep non slip soles
  • Adjustable shoe tie
  • Protection from biological agents, penetration of blood, body fluids and blood-borne pathogens
  • Anti-static: Tested according to EN 1149-5
Size: One size (fits size 42 - 46)
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge