OR6251T-10MG
OR6251T-5MG
APOSOR6251T-10MGEA
288
EUR
InStock
APOSOR6251T-10MG
APOSOR6251T-5MG
C2 Ceramide
C2 Ceramide
A biologically active, cell permeable, non-physiological ceramide analog. It inhibits cell proliferation, induces monocytic differentiation of HL-60 cells & inducesapoptosis.