B20033.03
B20033.06
B20033.14
B20033.03EA
49.1
EUR
InStock
B20033.03
B20033.06
B20033.14
4-(Trifluormethoxy)phenylisocyanat ≥97%
4-(Trifluormethoxy)phenylisocyanat
Purity: 97%
CAS No.: 35037-73-1
Molecular Formula: C8H4F3NO2
Formula Weight: 203.12
Storage Temperature: 2°C to 8°C
Flash Point: 70°(158°F)
Refractive Index: 1.4580
Melting Point: -29°
Boiling Point: 78°/22mm
Beilstein Reference: 977669
UN No.: 2206
MDL No.: MFCD00035702