B21458.03
B21458.06
B21458.14
B21458.03EA
61.5
EUR
InStock
B21458.03
B21458.06
B21458.14
5-Fluoranthranilsäure ≥98%
5-Fluoranthranilsäure
Purity: 98+%
CAS No.: 446-08-2
Molecular Formula: C7H6FNO2
Formula Weight: 155.13
Melting Point: 182-184°
Beilstein Reference: 2803664