B21992.0B
B21992.30
B21992.0BEA
69.5
EUR
InStock
B21992.0B
B21992.30
Polyethylenglycol 400
Polyethylenglycol 400
CAS No.: 25322-68-3
Molecular Formula: H(OCH2CH2)nOH
Flash Point: 240°(464°F)
Refractive Index: 1.4660
Odour: Odourless
pH: 5-7
Melting Point: 4-8°
Merck Reference: 14,7568
MDL No.: MFCD01779601