B22134.30
B22134.0B
B22134.30EA
24.8
EUR
InStock
B22134.30
B22134.0B
Polyethylenglycol 1.000
Polyethylenglycol 1.000
CAS No.: 25322-68-3
Molecular Formula: H(OCH2CH2)nOH
Flash Point: 260°(500°F)
Physical Form: Powder/low melting solid
Density: 1.101
Appearance: White
Odour: Odourless
pH: 5-7
Melting Point: 35-40°
Merck Reference: 14,7568
MDL No.: MFCD01779605